Tisk…

Bentonite, sodian

Dodavatel: Thermo Scientific

A15795.36 A15795.0E
A15795.36EA 906 CZK
A15795.36 A15795.0E
Bentonite, sodian
Bentonite, sodian

CAS No.: 1302-78-9 Physical Form: Powder Appearance: Cream to grey Odour: Odourless Merck Reference: 14,1055 MDL No.: MFCD00130611


Suspending agent, gel binder, filler in ceramics, catalyst support

Vzorec: Al₂H₂O₁₂Si₄
Hustota: 2,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00130611
CAS číslo: 1302-78-9
EINECS: 215-108-5
Merck Index: 14,1055

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR