Tisk…

1,4-Bis[α-(4-chlorophenyl)benzyl]piperazine dihydrochloride 97%

Dodavatel: Thermo Scientific

H63961.MD H63961.03 H63961.06
H63961.MDEA 1260 CZK
H63961.MD H63961.03 H63961.06
1,4-Bis[α-(4-chlorophenyl)benzyl]piperazine dihydrochloride 97%
1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride

Purity: 97% CAS No.: 346451-15-8 Molecular Formula: C30H28Cl2N2.2HCl Formula Weight: 560.38 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: White MDL No.: MFCD03844641

Vzorec: C₃₀H₃₀Cl₄N₂
MW: 560,384 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03844641
CAS číslo: 346451-15-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR