Tisk…

2-bromo-5-methylthiazole 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Bromo-5-methylthiazole

H63951.03 H63951.06
H63951.03EA 1140 CZK
H63951.03 H63951.06
2-bromo-5-methylthiazole 98%
2-bromo-5-methylthiazole

Purity: 98% CAS No.: 41731-23-1 Molecular Formula: C4H4BrNS Formula Weight: 178.05 MDL No.: MFCD08460610

Vzorec: C₄H₄BrNS
MW: 178,047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460610
CAS číslo: 41731-23-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR