Tisk…

N,N'-Dicarbobenzoxy-L-cystine 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: (Z-Cystine-OH)₂, (Z-Cys-OH)2, N,N'-Di-Cbz-L-cystine, N,N'-Dibenzyloxycarbonyl-L-cystine

H63917.06 H63917.14
H63917.06EA 1800 CZK
H63917.06 H63917.14
N,N'-Dicarbobenzoxy-L-cystine 98%
N,N'-Dicarbobenzoxy-L-cystine

Purity: 98% CAS No.: 6968-11-2 Molecular Formula: C22H24N2O8S2 Formula Weight: 508.56 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 119° Beilstein Reference: 2550640 MDL No.: MFCD00022030

Vzorec: C₂₂H₂₄N₂O₈S₂
MW: 508.56 g/mol
Bod tání: 119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00022030
CAS číslo: 6968-11-2
EINECS: 230-181-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR