Tisk…

2-Bromo-4,5-difluorophenylacetic acid 98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2-(2-Bromo-4,5-difluorophenyl)acetic acid

H63902.03 H63902.06
H63902.03EA 2880 CZK
H63902.03 H63902.06
2-Bromo-4,5-difluorophenylacetic acid 98%
2-Bromo-4,5-difluorophenylacetic acid

Purity: 98% CAS No.: 883502-07-6 Molecular Formula: C8H5BrF2O2 Formula Weight: 251.03 Physical Form: Solid Appearance: Off-white MDL No.: MFCD04973423

Vzorec: C₈H₅BrF₂O₂
MW: 251.027 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973423
CAS číslo: 883502-07-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR