Tisk…

N-(ɑ)-Boc-L-lysine 97%

Dodavatel: Thermo Fisher Scientific

Synonyms: (S)-6-Amino-2-((tert-butoxycarbonyl)amino)hexanoic acid, Nalpha-Boc-L-lysine, Boc-Lys-OH

H63859.06 H63859.14 H63859.22
H63859.06EA 3510 CZK
H63859.06 H63859.14 H63859.22
N-(ɑ)-Boc-L-lysine 97%
N-(alpha)-Boc-L-lysine

Purity: 97% CAS No.: 13734-28-6 Molecular Formula: C11H22N2O4 Formula Weight: 246.30 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: ca 205° dec Beilstein Reference: 4252546 Optical Rotation: +22°(c=2 in methanol) MDL No.: MFCD00038203

Vzorec: C₁₁H₂₂N₂O₄
MW: 246.3 g/mol
Bod tání: ∼ 205 °C (rozkl.)
MDL Number: MFCD00038203
CAS číslo: 13734-28-6
EINECS: 237-303-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR