4-(2-Fluorobenzyl)piperidine hydrochloride 97%
Dodavatel: Thermo Scientific
|
Varování
|
H63849.03
H63849.06
H63849.03EA
7760
CZK
H63849.03
H63849.06
4-(2-Fluorobenzyl)piperidine hydrochloride 97%
4-(2-Fluorobenzyl)piperidine hydrochloride
Purity: 97%
CAS No.: 193357-21-0
Molecular Formula: C12H16FN.HCl
Formula Weight: 229.72
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Off-white
MDL No.: MFCD09787475
Vzorec:
C₁₂H₁₇ClFN MW: 229,721 g/mol |
MDL Number:
MFCD09787475 CAS číslo: 193357-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...