1-[3-(Dimethylamino)propyl]-3-ethylurea 97%
Dodavatel: Thermo Scientific
|
Varování
|
H63845.MD
H63845.03
H63845.06
H63845.MDEA
1700
CZK
H63845.MD
H63845.03
H63845.06
1-[3-(Dimethylamino)propyl]-3-ethylurea 97%
1-[3-(Dimethylamino)propyl]-3-ethylurea
Purity: 97%
CAS No.: 32897-26-0
Molecular Formula: C8H19N3O
Formula Weight: 173.26
Beilstein Reference: 4961600
MDL No.: MFCD00027752
Vzorec:
C₈H₁₉N₃O Storage Temperature: Ambient |
MDL Number:
MFCD00027752 CAS číslo: 32897-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...