Tisk…

Fmoc-Arg(Boc)₂-OH 95%

Dodavatel: Thermo Fisher Scientific

Synonyms: Nomega,Nomega'-Di-Boc-Nalpha-Fmoc-L-arginine, Fmoc-Arg(Boc)2-OH

H63784.03 H63784.06
H63784.03EA 7690 CZK
H63784.03 H63784.06
Fmoc-Arg(Boc)₂-OH 95%
Fmoc-Arg(Boc)₂-OH

Purity: 95% CAS No.: 143824-77-5 Molecular Formula: C31H40N4O8 Formula Weight: 596.67 Physical Form: Solid MDL No.: MFCD00237008

Vzorec: C₃₁H₄₀N₄O₈
Storage Temperature: Ambient
MDL Number: MFCD00237008
CAS číslo: 143824-77-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR