L-Valinamide hydrochloride 95%
Dodavatel: Thermo Scientific
Synonyms:
H-Val-NH2.HCl, H-Val-NH₂·HCl
H63775.06
H63775.14
H63775.06EA
1540
CZK
H63775.06
H63775.14
L-Valinamide hydrochloride 95%
L-Valinamide hydrochloride
Purity: 95%
CAS No.: 3014-80-0
Molecular Formula: C5H12N2O.HCl
Formula Weight: 152.62
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 266-270°
Beilstein Reference: 4198583
Optical Rotation: +26°(c=1 in water)
MDL No.: MFCD00039085
Vzorec:
C₅H₁₂N₂O MW: 152,62 g/mol Bod tání: 266…270 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00039085 CAS číslo: 3014-80-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...