Tisk…

H-Lys(boc)-OMe·HCl 98%

Dodavatel: Thermo Fisher Scientific

H63727.03 H63727.06
H63727.03EA 2040 CZK
H63727.03 H63727.06
H-Lys(boc)-OMe·HCl 98%
H-Lys(boc)-OMe·HCl

Purity: 98% CAS No.: 2389-48-2 Molecular Formula: C12H24N2O4.HCl Formula Weight: 296.79 Storage Temperature: 2°C to 8°C Physical Form: Solid Beilstein Reference: 5173577 MDL No.: MFCD00076959

Vzorec: C₁₂H₂₅ClN₂O₄
MDL Number: MFCD00076959
CAS číslo: 2389-48-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR