Tisk…

Methyl-3-amino-5-methylthiophene-2-carboxylate 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: Methyl 3-amino-5-methylthiophene-2-carboxylate

H63719.03 H63719.06
H63719.03EA 5190 CZK
H63719.03 H63719.06
Methyl-3-amino-5-methylthiophene-2-carboxylate 97%
Methyl-3-amino-5-methylthiophene-2-carboxylate

Purity: 97% CAS No.: 76575-71-8 Molecular Formula: C7H9NO2S Formula Weight: 171.22 Physical Form: Solid MDL No.: MFCD00130095

Vzorec: C₇H₉NO₂S
MW: 171,214 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00130095
CAS číslo: 76575-71-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR