2,2,2-Trichloro-1-(1H-pyrrol-2-yl)ethanone 99+%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-(Trichloroacetyl)pyrrole, 2-(trichloroacetyl)-1h-pyrrole
H63718.06
H63718.14
H63718.06EA
1570
CZK
H63718.06
H63718.14
2,2,2-Trichloro-1-(1H-pyrrol-2-yl)ethanone 99+%
2,2,2-Trichloro-1-(1H-pyrrol-2-yl)ethanone
Purity: 99+%
CAS No.: 35302-72-8
Molecular Formula: C6H4Cl3NO
Formula Weight: 212.46
Physical Form: Powder
Appearance: Pale grey
Melting Point: 72-74°
Beilstein Reference: 135479
MDL No.: MFCD00128757
Vzorec:
C₆H₄Cl₃NO MW: 212,46 g/mol Bod tání: 72…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128757 CAS číslo: 35302-72-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...