Tisk…

H-Glu(OtBu)-OtBu·HCl 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: H-Glu(OtBu)-OtBu·HCl, L-Glutamic acid di-tert-butyl ester hydrochloride, H-Glu(OtBu)-OtBu.HCl

H63708.03 H63708.06 H63708.14
H63708.03EA 1760 CZK
H63708.03 H63708.06 H63708.14
H-Glu(OtBu)-OtBu·HCl 98%
H-Glu(OtBu)-OtBu·HCl

Purity: 98% CAS No.: 32677-01-3 Molecular Formula: C13H25NO4.HCl Formula Weight: 295.8 Storage Temperature: -30°C to -10°C Physical Form: Solid Beilstein Reference: 3573738 MDL No.: MFCD00058003

Vzorec: C₁₃H₂₆ClNO₄
MW: 295.804 g/mol
MDL Number: MFCD00058003
CAS číslo: 32677-01-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR