Tisk…

L-Proline-tert-butyl ester 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: (S)-tert-Butyl pyrrolidine-2-carboxylate, H-Pro-OtBu

H63657.03 H63657.06
H63657.03EA 1890 CZK
H63657.03 H63657.06
L-Proline-tert-butyl ester 98%
L-Proline-tert-butyl ester

Purity: 98% CAS No.: 2812-46-6 Molecular Formula: C9H17NO2 Formula Weight: 171.24 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Colourless MDL No.: MFCD00037879

Vzorec: C₉H₁₇NO₂
MW: 171,2 g/mol
MDL Number: MFCD00037879
CAS číslo: 2812-46-6
EINECS: 220-558-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR