Fmoc-Lys(Mtt)-OH 95%
Dodavatel: Thermo Scientific
Synonyms:
Nα-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-(4-methyltrityl)-L-lysine, Nα-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine
H63629.03
H63629.06
H63629.03EA
3400
CZK
H63629.03
H63629.06
Fmoc-Lys(Mtt)-OH 95%
Fmoc-Lys(Mtt)-OH
Purity: 95%
CAS No.: 167393-62-6
Molecular Formula: C41H40N2O4
Formula Weight: 624.77
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD00237166
Vzorec:
C₄₁H₄₀N₂O₄ MW: 624,781 g/mol |
MDL Number:
MFCD00237166 CAS číslo: 167393-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...