5-Amino-3-methyl-1-phenyl-1H-pyrazole 99+%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-Methyl-1-phenyl-1H-pyrazol-5-amine, 5-Amino-3-methyl-1-phenylpyrazole, 3-methyl-1-phenyl-1H-pyrazol-5-amine
H63605.06
H63605.06EA
1780
CZK
H63605.06
5-Amino-3-methyl-1-phenyl-1H-pyrazole 99+%
5-Amino-3-methyl-1-phenyl-1H-pyrazole
Purity: 99+%
CAS No.: 1131-18-6
Molecular Formula: C10H11N3
Formula Weight: 173.21
Physical Form: Solid
Appearance: Yellow to pale brown
Melting Point: 114-117°
Beilstein Reference: 140666
MDL No.: MFCD00020727
Vzorec:
C₁₀H₁₁N₃ MW: 173,21 g/mol Bod tání: 114…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020727 CAS číslo: 1131-18-6 EINECS: 214-463-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...