N-Methylpyridin-3-amine 95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
3-(Methylamino)pyridine
H63602.MD
H63602.03
H63602.06
H63602.MDEA
1460
CZK
H63602.MD
H63602.03
H63602.06
N-Methylpyridin-3-amine 95%
N-Methylpyridin-3-amine
Purity: 95%
CAS No.: 18364-47-1
Molecular Formula: C6H8N2
Formula Weight: 108.14
Physical Form: Liquid
Appearance: Yellow
UN No.: UN2810
MDL No.: MFCD01418130
Vzorec:
C₆H₈N₂ Storage Temperature: Ambient |
MDL Number:
MFCD01418130 CAS číslo: 18364-47-1 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...