Fmoc-Chg-OH 98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Fmoc-L-Chg-OH, Fmoc-cyclohexyl-Gly-OH, N-Fmoc-L-cyclohexylglycine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-cyclohexylglycine, N-Fmoc-2-cyclohexyl-L-glycine
H63589.MD
H63589.03
H63589.06
H63589.MDEA
1180
CZK
H63589.MD
H63589.03
H63589.06
Fmoc-Chg-OH 98%
Fmoc-Chg-OH
Purity: 98%
CAS No.: 161321-36-4
Molecular Formula: C23H25NO4
Formula Weight: 379.45
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD00190878
Vzorec:
C₂₃H₂₅NO₄ |
MDL Number:
MFCD00190878 CAS číslo: 161321-36-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...