Tisk…

Fmoc-Asp(OAll)-OH 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: Fmoc-Asp(allyl ester)-OH, N-Fmoc-L-aspartic acid 4-allyl ester

H63585.06 H63585.14
H63585.06EA 3440 CZK
H63585.06 H63585.14
Fmoc-Asp(OAll)-OH 98%
Fmoc-Asp(OAll)-OH

Purity: 98% CAS No.: 146982-24-3 Molecular Formula: C22H21NO6 Formula Weight: 395.41 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 111-115° Beilstein Reference: 6670219 Optical Rotation: -27°(c=1 in DMF) MDL No.: MFCD00190874

Vzorec: C₂₂H₂₁NO₆
Bod tání: 111…115 °C
MDL Number: MFCD00190874
CAS číslo: 146982-24-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR