Tisk…

6-Chloro-4-methylpyridine-2-carboxylic acid 95%

Dodavatel: Thermo Scientific
Nebezpečí

H63584.03 H63584.06
H63584.03EA 4020 CZK
H63584.03 H63584.06
6-Chloro-4-methylpyridine-2-carboxylic acid 95%
6-Chloro-4-methylpyridine-2-carboxylic acid

Purity: 95% CAS No.: 324028-95-7 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Physical Form: Solid Appearance: Off-white UN No.: UN2811 MDL No.: MFCD09834827

Vzorec: C₇H₆ClNO₂
MW: 171,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834827
CAS číslo: 324028-95-7
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR