Boc-4-(Fmoc-amino)-1-phenylalanine 95%
Dodavatel: Thermo Fisher Scientific
Synonyms:
(S)-3-(4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid, Boc-p-amino-Phe(Fmoc)-OH, Boc-Phe(4-NHFmoc)-OH, N-Boc-4-(Fmoc-amino)-L-phenylalanine
H63543.03
H63543.06
H63543.03EA
2030
CZK
H63543.03
H63543.06
Boc-4-(Fmoc-amino)-1-phenylalanine 95%
Boc-4-(Fmoc-amino)-1-phenylalanine
Purity: 95%
CAS No.: 114346-31-5
Molecular Formula: C29H30N2O6
Formula Weight: 502.56
Physical Form: Solid
MDL No.: MFCD00151889
Vzorec:
C₂₉H₃₀N₂O₆ MW: 502.567 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00151889 CAS číslo: 114346-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...