N-Formyl-L-methionine 95%
Dodavatel: Thermo Fisher Scientific
Synonyms:
For-Met-OH, Formyl-L-methionine
H63493.MD
H63493.03
H63493.06
H63493.MDEA
1030
CZK
H63493.MD
H63493.03
H63493.06
N-Formyl-L-methionine 95%
N-Formyl-L-methionine
Purity: 95%
CAS No.: 4289-98-9
Molecular Formula: C6H11NO3S
Formula Weight: 177.22
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD00021033
Vzorec:
C₆H₁₁NO₃S |
MDL Number:
MFCD00021033 CAS číslo: 4289-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...