N-[(9H-Fluoren-9-ylmethoxy)carbonyl]anthranilic acid 98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Fmoc-2-Abz-OH, 2-(Fmoc-amino)benzoic Acid, 2-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]benzoic Acid, N-Fmoc-anthranilic Acid
H63459.06
H63459.14
H63459.06EA
2260
CZK
H63459.06
H63459.14
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]anthranilic acid 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]anthranilic acid
Purity: 98%
CAS No.: 150256-42-1
Molecular Formula: C22H17NO4
Formula Weight: 359.37
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 216-218° dec
Beilstein Reference: 7659726
MDL No.: MFCD00235883
Vzorec:
C₂₂H₁₇NO₄ |
MDL Number:
MFCD00235883 CAS číslo: 150256-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...