Fmoc-4-chloro-L-phenylalanine 98%
Dodavatel: Thermo Scientific
Synonyms:
Fmoc-4-chloro-Phe-OH, N-Fmoc-4-chloro-L-phenylalanine, Fmoc-Phe(4-Cl)-OH
H63448.03
H63448.06
H63448.03EA
3210
CZK
H63448.03
H63448.06
Fmoc-4-chloro-L-phenylalanine 98%
Fmoc-4-chloro-L-phenylalanine
Purity: 98%
CAS No.: 175453-08-4
Molecular Formula: C24H20ClNO4
Formula Weight: 421.87
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 9437838
MDL No.: MFCD00080281
Vzorec:
C₂₄H₂₀ClNO₄ MW: 421,877 g/mol |
MDL Number:
MFCD00080281 CAS číslo: 175453-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...