Tisk…

Ethyl-4-nitro-1H-indole-2-carboxylate 97%

Dodavatel: Thermo Scientific

Synonyms: Ethyl 4-nitroindole-2-carboxylate, Ethyl 4-nitro-1H-indole-2-carboxylate

H63440.MD H63440.03
H63440.MDEA 7450 CZK
H63440.MD H63440.03
Ethyl-4-nitro-1H-indole-2-carboxylate 97%
Ethyl-4-nitro-1H-indole-2-carboxylate

Purity: 97% CAS No.: 4993-93-5 Molecular Formula: C11H10N2O4 Formula Weight: 234.21 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: Yellow MDL No.: MFCD06795512

Vzorec: C₁₁H₁₀N₂O₄
MW: 234,211 g/mol
MDL Number: MFCD06795512
CAS číslo: 4993-93-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR