N-Benzoyl-L-tyrosine ethyl ester 98%
Dodavatel: Thermo Scientific
Synonyms:
BTEE, Bz-Tyr-OEt
H63434.06
H63434.14
H63434.06EA
1640
CZK
H63434.06
H63434.14
N-Benzoyl-L-tyrosine ethyl ester 98%
N-Benzoyl-L-tyrosine ethyl ester
Purity: 98%
CAS No.: 3483-82-7
Molecular Formula: C18H19NO4
Formula Weight: 313.35
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point: 118-121°
Beilstein Reference: 2223819
MDL No.: MFCD00002388
Vzorec:
C₁₈H₁₉NO₄ MW: 313,35 g/mol Bod tání: 118…121 °C Storage Temperature: Freezer |
MDL Number:
MFCD00002388 CAS číslo: 3483-82-7 EINECS: 222-469-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...