Tisk…

4-(Trifluoromethylthio)phenylacetic acid 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-(4-((Trifluoromethyl)thio)phenyl)acetic acid, 4-(Carboxymethyl)phenyl trifluoromethyl sulphide, {4-[(Trifluoromethyl)sulphanyl]phenyl}acetic acid, 4-[(Trifluoromethyl)sulphanyl]phenylacetic acid

H63432.03 H63432.06
H63432.03EA 2090 CZK
H63432.03 H63432.06
4-(Trifluoromethylthio)phenylacetic acid 98%
4-(Trifluoromethylthio)phenylacetic acid

Purity: 98% CAS No.: 243977-23-3 Molecular Formula: C9H7F3O2S Formula Weight: 236.21 Physical Form: Solid MDL No.: MFCD00236351

Vzorec: C₉H₇F₃O₂S
MW: 236,208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236351
CAS číslo: 243977-23-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR