Boc-O-benzyl-D-tyrosine 95%
Dodavatel: Thermo Fisher Scientific
Synonyms:
N-Boc-O-benzyl-D-tyrosine, Boc-D-Tyr(Bzl)-OH
H63427.06
H63427.14
H63427.06EA
3210
CZK
H63427.06
H63427.14
Boc-O-benzyl-D-tyrosine 95%
Boc-O-benzyl-D-tyrosine
Purity: 95%
CAS No.: 63769-58-4
Molecular Formula: C21H25NO5
Formula Weight: 371.43
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD00038249
Vzorec:
C₂₁H₂₅NO₅ MW: 371.433 g/mol |
MDL Number:
MFCD00038249 CAS číslo: 63769-58-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...