D-Methionine methyl ester hydrochloride 98%
Dodavatel: Thermo Fisher Scientific
H63422.06
H63422.14
H63422.22
H63422.06EA
1120
CZK
H63422.06
H63422.14
H63422.22
D-Methionine methyl ester hydrochloride 98%
D-Methionine methyl ester hydrochloride
Purity: 98%
CAS No.: 69630-60-0
Molecular Formula: C6H13NO2S.HCl
Formula Weight: 199.7
Physical Form: Solid
MDL No.: MFCD00070385
Vzorec:
C₆H₁₃NO₂S·HCl Storage Temperature: Ambient |
MDL Number:
MFCD00070385 CAS číslo: 69630-60-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...