3-(2-Naphthyl)-L-alanine 97%
Dodavatel: Thermo Scientific
Synonyms:
(S)-alpha-Amino-2-naphthalenepropionic Acid, (S)-2-Amino-3-(naphthalen-2-yl)propanoic acid, H-3-(2-Naphthyl)-Ala-OH
Purity: 97%
CAS No.: 58438-03-2
Molecular Formula: C13H13NO2
Formula Weight: 215.25
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 2726272
Optical Rotation: -11°(c= 1M HCl)
MDL No.: MFCD00066087
Vzorec:
C₁₃H₁₃NO₂ MW: 215,25 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00066087 CAS číslo: 58438-03-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...