Thiazole-2-carbaldehyde 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Thiazolecarboxaldehyde, Thiazole-2-carboxaldehyde
H63329.03
H63329.06
H63329.03EA
1960
CZK
H63329.03
H63329.06
Thiazole-2-carbaldehyde 95%
Thiazole-2-carbaldehyde
Purity: 95%
CAS No.: 10200-59-6
Molecular Formula: C4H3NOS
Formula Weight: 113.13
Storage Temperature: 2°C to 8°C
Flash Point: 68°(154°F)
Physical Form: Liquid
Density: 1.288
Refractive Index: 1.574
Appearance: Colourless to pale yellow
Boiling Point: 61-63°/15mm
MDL No.: MFCD00142924
Vzorec:
C₄H₃NOS MW: 113,13 g/mol Bod varu: 61…63 °C (15 mmHg) Hustota: 1,288 Bod vzplanutí: 68 °C (154 °F) |
MDL Number:
MFCD00142924 CAS číslo: 10200-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...