Tisk…

4-[2-(Boc-amino)ethyl]aniline 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: tert-Butyl 4-aminophenethylcarbamate, 4-(2-Aminoethyl)aniline, 4-BOC protected

H63324.03 H63324.06
H63324.03EA 3920 CZK
H63324.03 H63324.06
4-[2-(Boc-amino)ethyl]aniline 97%
4-[2-(Boc-amino)ethyl]aniline

Purity: 97% CAS No.: 94838-59-2 Molecular Formula: C13H20N2O2 Formula Weight: 236.31 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: Brown UN No.: UN2811 MDL No.: MFCD05663961

Vzorec: C₁₃H₂₀N₂O₂
MW: 236,315 g/mol
MDL Number: MFCD05663961
CAS číslo: 94838-59-2
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR