Tisk…

h-d-Tyr-otbu 98%

Dodavatel: Thermo Scientific

Synonyms: H-D-Tyr-OtBu, D-Tyrosine tert-butyl ester

H63274.03 H63274.06
H63274.03EA 1250 CZK
H63274.03 H63274.06
h-d-Tyr-otbu 98%
h-d-Tyr-otbu

Purity: 98% CAS No.: 87553-74-0 Molecular Formula: C13H19NO3 Formula Weight: 237.14 Storage Temperature: 2°C to 8°C Physical Form: Solid MDL No.: MFCD00191035

Vzorec: C₁₃H₁₉NO₃
MW: 237,299 g/mol
MDL Number: MFCD00191035
CAS číslo: 87553-74-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR