N-Carbobenzoxy-L-valine succinimidyl ester 98%
Dodavatel: Thermo Scientific
Synonyms:
N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, (S)-2,5-Dioxopyrrolidin-1-yl 2-(((benzyloxy)carbonyl)amino)-3-methylbutanoate, N-Cbz-L-valine Succinimidyl Ester, Z-Val-OSu
H63253.06
H63253.14
H63253.06EA
1580
CZK
H63253.06
H63253.14
N-Carbobenzoxy-L-valine succinimidyl ester 98%
N-Carbobenzoxy-L-valine succinimidyl ester
Purity: 98%
CAS No.: 3496-11-5
Molecular Formula: C17H20N2O6
Formula Weight: 348.35
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 114-118°
Beilstein Reference: 1555287
Optical Rotation: -25°(c=1 in dioxane)
MDL No.: MFCD00053547
Vzorec:
C₁₇H₂₀N₂O₆ MW: 348,36 g/mol Bod tání: 114…118 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00053547 CAS číslo: 3496-11-5 EINECS: 222-493-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...