Tisk…

1-Methyl-L-aspartate 98%

Dodavatel: Thermo Fisher Scientific

H63221.03 H63221.06
H63221.03EA 1490 CZK
H63221.03 H63221.06
1-Methyl-L-aspartate 98%
1-Methyl-L-aspartate

Purity: 98% CAS No.: 17812-32-7 Molecular Formula: C5H9NO4 Formula Weight: 147.13 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 186° dec Beilstein Reference: 2080763 Optical Rotation: +36°(c=0.5 in water) MDL No.: MFCD00080289

Vzorec: C₅H₉NO₄
MDL Number: MFCD00080289
CAS číslo: 17812-32-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR