Fmoc-Lys(Dabcyl)-OH 95%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Nepsilon-4-[4-(Dimethylamino)phenylazo]benzoyl-Nα-Fmoc-L-lysine
H63214.MD
H63214.03
H63214.MDEA
4770
CZK
H63214.MD
H63214.03
Fmoc-Lys(Dabcyl)-OH 95%
Fmoc-Lys(Dabcyl)-OH
Purity: 95%
CAS No.: 146998-27-8
Molecular Formula: C36H37N5O5
Formula Weight: 619.71
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: Red
MDL No.: MFCD00467694
Vzorec:
C₃₆H₃₇N₅O₅ MW: 619.722 g/mol |
MDL Number:
MFCD00467694 CAS číslo: 146998-27-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...