Tisk…

Ethyl-2-methylquinoline-6-carboxylate 97%

Dodavatel: Thermo Scientific

Synonyms: Ethyl 2-methylquinoline-6-carboxylate

H63213.03 H63213.06
H63213.03EA 2480 CZK
H63213.03 H63213.06
Ethyl-2-methylquinoline-6-carboxylate 97%
Ethyl-2-methylquinoline-6-carboxylate

Purity: 97% CAS No.: 855763-77-8 Molecular Formula: C13H13NO2 Formula Weight: 215.25 Physical Form: Solid MDL No.: MFCD09879697

Vzorec: C₁₃H₁₃NO₂
MW: 215,252 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09879697
CAS číslo: 855763-77-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR