N-Boc-guanidine 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Aminoiminomethyl)carbamic acid tert-butyl ester, tert-Butoxycarbonylguanidine
H63206.03
H63206.06
H63206.14
H63206.03EA
656
CZK
H63206.03
H63206.06
H63206.14
N-Boc-guanidine 95%
N-Boc-guanidine
N-Boc-guanidine, is used as used as guanidinylating agent and for synthesis of 2-aminoimidazoles. It is also used as a pharmaceutical intermediate.
Purity: 95%
CAS No.: 219511-71-4
Molecular Formula: C6H13N3O2
Formula Weight: 159.19
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 170-175°
MDL No.: MFCD20547465
Vzorec:
C₆H₁₃N₃O₂ MW: 159,189 g/mol Bod tání: 170…175 °C |
MDL Number:
MFCD20547465 CAS číslo: 219511-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...