(R)-Dimethyl-2-aminosuccinate hydrochloride 98%
Dodavatel: Thermo Scientific
Synonyms:
H-D-Asp(OMe)-OMe.HCl, D-Aspartic acid dimethyl ester hydrochloride, H-D-Asp(OMe)-OMe·HCl
Purity: 98%
CAS No.: 69630-50-8
Molecular Formula: C6H11NO4.HCl
Formula Weight: 197.62
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point: 114-119°
Optical Rotation: -14°(c=1 in methanol)
MDL No.: MFCD00070384
Vzorec:
C₆H₁₂ClNO₄ MW: 197,615 g/mol Bod tání: 114…119 °C |
MDL Number:
MFCD00070384 CAS číslo: 69630-50-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...