Tisk…

3,4-Xylylacetic acid 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3,4-Dimethylphenylacetic acid

H63178.03 H63178.06
H63178.03EA 1530 CZK
H63178.03 H63178.06
3,4-Xylylacetic acid 98%
3,4-Xylylacetic acid

Purity: 98% CAS No.: 17283-16-8 Molecular Formula: C10H12O2 Formula Weight: 164.2 Physical Form: Solid MDL No.: MFCD02664684

Vzorec: C₁₀H₁₂O₂
MW: 164,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664684
CAS číslo: 17283-16-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR