Tisk…

N-Carbobenzoxy-D-methionine 98%

Dodavatel: Thermo Fisher Scientific

H63152.06 H63152.14
H63152.06EA 1010 CZK
H63152.06 H63152.14
N-Carbobenzoxy-D-methionine 98%
N-Carbobenzoxy-D-methionine

Purity: 98% CAS No.: 28862-80-8 Molecular Formula: C13H17NO4S Formula Weight: 283.24 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 68-72° dec Beilstein Reference: 2058695 Optical Rotation: +26°(c=1 in methanol) MDL No.: MFCD00026043

Vzorec: C₁₃H₁₇NO₄S
Storage Temperature: Refrigerator
MDL Number: MFCD00026043
CAS číslo: 28862-80-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR