Tisk…

Ethyl-1-boc-piperazine-3-carboxylate 97%

Dodavatel: Thermo Scientific
Varování

H63146.03 H63146.06
H63146.03EA 7110 CZK
H63146.03 H63146.06
Ethyl-1-boc-piperazine-3-carboxylate 97%
Ethyl-1-boc-piperazine-3-carboxylate

Purity: 97% CAS No.: 183742-29-2 Molecular Formula: C12H22N2O4 Formula Weight: 258.31 Physical Form: Solid MDL No.: MFCD13172266

Vzorec: C₁₂H₂₂N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD13172266
CAS číslo: 183742-29-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR