3-(3-Pyridyl)-D-alanine 95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H63132.03
H63132.06
H63132.03EA
4290
CZK
H63132.03
H63132.06
3-(3-Pyridyl)-D-alanine 95%
3-(3-Pyridyl)-D-alanine
Purity: 95%
CAS No.: 70702-47-5
Molecular Formula: C8H10N2O2
Formula Weight: 166.18
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Off-white
Beilstein Reference: 4351584
MDL No.: MFCD00079673
Vzorec:
C₈H₁₀N₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00079673 CAS číslo: 70702-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...