Tisk…

3-Fluoro-D-phenylalanine 95%

Dodavatel: Thermo Fisher Scientific

H63131.03 H63131.06
H63131.03EA 2460 CZK
H63131.03 H63131.06
3-Fluoro-D-phenylalanine 95%
3-Fluoro-D-phenylalanine

Purity: 95% CAS No.: 110117-84-5 Molecular Formula: C9H10FNO2 Formula Weight: 183.18 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD00066449

Vzorec: C₉H₁₀FNO₂
MDL Number: MFCD00066449
CAS číslo: 110117-84-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR