DL-Homoserine 98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
H-DL-HoSer-OH, H-DL-Hser-OH, H-DL-HomoSer-OH, (±)-Homoserine
H63118.03
H63118.06
H63118.03EA
2490
CZK
H63118.03
H63118.06
DL-Homoserine 98%
DL-Homoserine
Purity: 98%
CAS No.: 1927-25-9
Molecular Formula: C4H9NO3
Formula Weight: 119.12
Physical Form: Solid
Melting Point: 188-189°
Beilstein Reference: 1721682
MDL No.: MFCD00002618
Vzorec:
C₄H₉NO₃ Bod tání: 188…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002618 CAS číslo: 1927-25-9 EINECS: 217-661-8 Merck Index: 14,4739 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...