2-Cyano-5-fluorobenzyl bromide 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H63110.06
H63110.14
H63110.06EA
4090
CZK
H63110.06
H63110.14
2-Cyano-5-fluorobenzyl bromide 98%
2-Cyano-5-fluorobenzyl bromide
Purity: 98%
CAS No.: 421552-12-7
Molecular Formula: C8H5BrFN
Formula Weight: 214.04
Physical Form: Solid
UN No.: UN2923
MDL No.: MFCD08059542
Vzorec:
C₈H₅BrFN Storage Temperature: Ambient |
MDL Number:
MFCD08059542 CAS číslo: 421552-12-7 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...