3,5-Di-tert-butyl-4-hydroxycinnamic acid predominantly trans 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A10514.09
A10514.18
A10514.09EA
1920
CZK
A10514.09
A10514.18
3,5-Di-tert-butyl-4-hydroxycinnamic acid predominantly trans 98%
3,5-Di-tert-butyl-4-hydroxycinnamic acid
Purity: 98%
CAS No.: 2243-42-7
Molecular Formula: C13H10O3
Formula Weight: 214.22
Melting Point: 111-115°
Beilstein Reference: 978452
Vzorec:
C₁₇H₂₄O₃ Bod varu: 355 °C (1013 hPa) Hustota: 1,2141 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00017291 CAS číslo: 22014-01-3 EINECS: 218-811-5 |
Výsledky specifikačního testu
Appearance (Colour) | White to cream or pale yellow or pale orange |
Form | Powder |
Assay (Aqueous acidbase Titration) | >97.5 to <102.5% |
Assay (Silylated GC) | >97.5% (sum of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...