DL-Glutamic acid 95%
Dodavatel: Thermo Scientific
H63075.14
H63075.22
H63075.36
H63075.14EA
2390
CZK
H63075.14
H63075.22
H63075.36
DL-Glutamic acid 95%
DL-Glutamic acid
Purity: 95%
CAS No.: 617-65-2
Molecular Formula: C5H9NO4
Formula Weight: 147.13
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 1723799
MDL No.: MFCD00063113
Vzorec:
C₅H₉NO₄ MW: 147,13 g/mol |
MDL Number:
MFCD00063113 CAS číslo: 617-65-2 EINECS: 210-522-2 Merck Index: 14,4469 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...