Tisk…

O-tert-Butyl-L-threonine 97%

Dodavatel: Thermo Fisher Scientific

H63031.06 H63031.14
H63031.06EA 2870 CZK
H63031.06 H63031.14
O-tert-Butyl-L-threonine 97%
O-tert-Butyl-L-threonine

Purity: 97% CAS No.: 4378-13-6 Molecular Formula: C8H17NO3 Formula Weight: 175.23 Physical Form: Solid Beilstein Reference: 2206127 Optical Rotation: -43°(c=1 in water) MDL No.: MFCD00037766

Vzorec: C₈H₁₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD00037766
CAS číslo: 4378-13-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR